EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Quantification methods of folpet degradation products in plasma with HPLC-UV/DAD: application to an in vivo toxicokinetic study in rats.
PMID 18343208
Abstract
Solid-phase extractions followed by HPLC-UV/DAD methods were developed for occupational biological monitoring or forensic investigations of the fungicide folpet using its degradation products, phthalimide and phthalamic acid as plasma biomarkers. These methods show good linearity (r>0.9955), precision (CV<15%) and accuracy (bias<14.8%). The lower limits of quantification for phthalimide and phthalamic acid were 10 and 20 ng/ml and the absolute recoveries were higher than 86% and 68%, respectively. Applying these methods, a plasma toxicokinetic study of folpet in rats after intratracheal administration of Folpan 80WG showed that inhalation of folpet could be a route of exposure with an important systemic absorption.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32057
Folpet, PESTANAL®, analytical standard
C9H4Cl3NO2S