EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
Acute oculogyric crisis in a patient taking clebopride.
PMID 18344644
Abstract
We present a 21-year-old woman with an oculogyric crisis following the administration of clebopride. Based on the temporal relationship of the administration of clebopride to the occurrence of the event and the absence of other possible etiologies, clebopride is implicated as the cause of this acute oculogyric crisis, despite the fact that a cause-and-effect relationship cannot be confirmed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8289
Clebopride maleate salt, ~95%
C20H24ClN3O2 · C4H4O4