EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Phosphorylation of the androgen receptor is associated with reduced survival in hormone-refractory prostate cancer patients.
PMID 18349820
Abstract
Cell line studies demonstrate that the PI3K/Akt pathway is upregulated in hormone-refractory prostate cancer (HRPC) and can result in phosphorylation of the androgen receptor (AR). The current study therefore aims to establish if this has relevance to the development of clinical HRPC. Immunohistochemistry was employed to investigate the expression and phosphorylation status of Akt and AR in matched hormone-sensitive and -refractory prostate cancer tumours from 68 patients. In the hormone-refractory tissue, only phosphorylated AR (pAR) was associated with shorter time to death from relapse (P=0.003). However, when an increase in expression in the transition from hormone-sensitive to -refractory prostate cancer was investigated, an increase in expression of PI3K was associated with decreased time to biochemical relapse (P=0.014), and an increase in expression of pAkt(473) and pAR(210) were associated with decreased disease-specific survival (P=0.0019 and 0.0015, respectively). Protein expression of pAkt(473) and pAR(210) also strongly correlated (P<0.001, c.c.=0.711) in the hormone-refractory prostate tumours. These results provide evidence using clinical specimens, that upregulation of the PI3K/Akt pathway is associated with phosphorylation of the AR during development of HRPC, suggesting that this pathway could be a potential therapeutic target.