EMAIL THIS PAGE TO A FRIEND
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine
[Protective effect of 3-hydroxypyridine derivatives in extreme conditions].
PMID 18350829
Abstract
The paper presents the results of experimental and clinical testing of 3-hydroxypyridine derivatives (emoxipin, mexidol and new compounds) various extreme conditions (hypoxia, hyper- and hypothermia, vibration, hypokinesia, ionizing and nonionizing radiation and others). Analysis of the results led to the conclusion that 3-hydroxipiridine derivatives are a group of compounds worthy of further in-depth investigations in a broad range of extreme conditions in order to assess their potential use in development of new medical protectors of humans and animals facing extreme factors of various nature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H57009
3-Hydroxypyridine, 98%
C5H5NO