EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
A stochastic model for predicting dextrose equivalent and saccharide composition during hydrolysis of starch by alpha-amylase.
PMID 18351657
Abstract
A stochastic model was developed that was used to describe the formation and breakdown of all saccharides involved during alpha-amylolytic starch hydrolysis in time. This model is based on the subsite maps found in literature for Bacillus amyloliquefaciens alpha-amylase (BAA) and Bacillus licheniformis alpha-amylase (BLA). Carbohydrate substrates were modeled in a relatively simple two-dimensional matrix. The predicted weight fractions of carbohydrates ranging from glucose to heptasaccharides and the predicted dextrose equivalent showed the same trend and order of magnitude as the corresponding experimental values. However, the absolute values were not the same. In case a well-defined substrate such as maltohexaose was used, comparable differences between the experimental and simulated data were observed indicating that the substrate model for starch does not cause these deviations. After changing the subsite map of BLA and the ratio between the time required for a productive and a non-productive attack for BAA, a better agreement between the model data and the experimental data was observed. Although the model input should be improved for more accurate predictions, the model can already be used to gain knowledge about the concentrations of all carbohydrates during hydrolysis with an alpha-amylase. In addition, this model also seems to be applicable to other depolymerase-based systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9153
Maltohexaose, ≥65% (HPLC)
C36H62O31