EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Lithium thiazolidine-4-carboxylate: synthesis, spectroscopic characterization and preliminary in vitro cytotoxic studies in human HeLa cells.
PMID 18353709
Abstract
A new water-soluble lithium salt of thiazolidine-4-carboxylic acid was synthesized and characterized by chemical and spectroscopic techniques. Elemental and mass spectrometric (ESI-MS) analyses of the solid compound fit to the composition LiC(4)H(6)NSO(2). (1)H, (13)C nuclear magnetic resonance (NMR), [(1)H-(15)N] NMR and infrared (IR) analyses permitted to elucidate the structure of the compound. Biological activity was evaluated by cytotoxic analysis using HeLa cells. Determination of cell death was assessed using a tetrazolium salt colorimetric assay, which reflects the cells viability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T27502
L-4-Thiazolidinecarboxylic acid, 98%
C4H7NO2S