EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Vibrational spectroscopic studies and ab initio calculations of a substituted amide of pyrazine-2-carboxylic acid--C12H10ClN3O.
PMID 18353710
Abstract
A substituted amide of pyrazine-2-carboxylic acid was prepared and the IR spectrum is recorded and analysed. The vibrational frequencies and corresponding vibrational assignments are examined theoretically using the Gaussian03 set of quantum chemistry codes. Predicted infrared and Raman intensities are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P56100
Pyrazinecarboxylic acid, 99%
C5H4N2O2