EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
TIM-4 expressed on APCs induces T cell expansion and survival.
PMID 18354194
Abstract
TIM (T cell, Ig, mucin) proteins can regulate T cell immune responses. Tim-4 mRNA is not expressed in T cells, but exclusively in APCs. Tim-4 is a ligand for Tim-1 and Tim-4.Ig fusion protein was shown to either inhibit or expand T cells. However, the molecular basis for such opposite effects was not defined. By generating mAbs, we show that expression of Tim-4 protein is restricted to CD11c(+) and CD11b(+) cells and is up-regulated upon activation. We show that Tim-4 specifically phosphorylates Tim-1 and induces T cell expansion by enhancing cell division and reducing apoptosis. Tim-4 also induces the phosphorylation of signaling molecules LAT, Akt, and ERK1/2 in T cells. Tim-4, expressed on APCs, is a costimulatory molecule that promotes T cell expansion and survival by cross-linking Tim-1 on T cells.