EMAIL THIS PAGE TO A FRIEND
Ukrains'kyi biokhimichnyi zhurnal (1999 )
[Lectins from Sambucus nigra L inflorescences: isolation and investigation of biological activity using procaryotic test-systems].
PMID 18357788
Abstract
Isolation of lectins from extracts of the Sambucus nigra inflorescences and of pollen material have been performed using isoelectric focusing without carrier ampholytes (autofocusing). Fractions active in agglutination tests with different carbohydrate specificity were subjected to SDS-PAGE. The major lectin found in whole inflores-cences was GalNAc specific and is proposed to be a heterotetramer with subunits of about 30 and 33 kDa. It was called SNAflu-I. At least two other lectins were present in the pollen material and supposed to consist of identical subunits. Major positively charged lectin was Glc/Man specific with subunit of 26 kDa and called SNApol-I. Other pollen component (SNApol-II) was Gal specific with subunit of about 20 kDa. In order to elucidate cell targets sensitive for the S. nigra lectin's activity the combined effects of the lectins and transcriptional of phenazine origin on B. subtilis cells growth have been studied. Only SNApol-I demonstrated the antagonistic activity against these inhibitors in vivo. This lectin but not the SNAflu-I can also inhibit transcription in vitro. It is supposed that lectins from the same source may act in different directions on cell metabolism. Particularly one of the common targets may be the DNA-dependent synthesis of RNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1757
6-Azauracil, ≥98%
C3H3N3O2