EMAIL THIS PAGE TO A FRIEND
The Journal of arthroplasty
"A randomized clinical trial of cementless femoral stems with and without hydroxyapatite/tricalcium-phosphate coating: an 8- to 12-year follow-up study" by Yoon et al.
PMID 18358396
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7263
Ethynodiol diacetate
C24H32O4