EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Rhodamine-based fluorogenic probe for imaging biological thiol.
PMID 18358719
Abstract
We have developed a new fluorescent probe for biological thiol. The probe was synthesized by the modification of the 2,4-dinitrobenzenesulfonyl group with rhodamine 110. The selective detection of thiol species such as cysteine or glutathione was achieved in biological conditions. Moreover, the probe was successfully applied to the imaging of thiol species in living human cells.
Related Materials