EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Directed evolution of a maltogenic alpha-amylase from Bacillus sp. TS-25.
PMID 18359119
Abstract
Directed evolution coupled with a high-throughput robotic screen was employed to broaden the industrial use of the maltogenic alpha-amylase Novamyl from Bacillus sp. TS-25. Wild-type Novamyl is currently used in the baking industry as an anti-staling agent in breads baked at neutral or near neutral pH. However, the enzyme is rapidly inactivated during the baking process of bread made with low pH recipes and Novamyl thus has very limited beneficial effect for this particular application. In an effort to improve the performance of Novamyl for low pH bread applications such as sourdough and rye, two error-prone PCR libraries were generated, expressed in Bacillus subtilis and screened for variants with improved thermal stability and activity under low pH conditions. Variants exhibiting improved performance were iteratively recombined using DNA shuffling to create two generations of libraries. Relative to wild-type Novamyl, a number of the resulting variants exhibited more than 10 degrees C increase in thermal stability at pH 4.5, one of which demonstrated substantial anti-staling properties in low pH breads.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2986
Amylase, Maltogenic from Bacillus sp.