Description
Molecular Formula
Add to Cart
1286413
Frovatriptan succinate, United States Pharmacopeia (USP) Reference Standard
C14H17N3O · C4H6O4 · H2O
SML1291
Frovatriptan succinate monohydrate, ≥97% (HPLC)
C14H17N3O · C4H6O4 · H2O