EMAIL THIS PAGE TO A FRIEND
Molecular diversity
Reaction of 5-aminotetrazole with vinamidinium salts: formation of 2-(N, N-dimethylamino)-5-substituted pyrimidines.
PMID 18365762
Abstract
The cyclocondensation of vinamidinium salts with 5-aminotetrazole in basic media affords regiospecifically good yields of 2-(N, N-dimethylamino)-5- aryl-, arylsulfonyl- and 2-(N, N-dimethylamino)-5-formyl-pyrimidines, respectively.