EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Protein scaffold of a designed metalloenzyme enhances the chemoselectivity in sulfoxidation of thioanisole.
PMID 18368158
Abstract
We demonstrate that incorporation of MnSalen into a protein scaffold enhances the chemoselectivity in sulfoxidation of thioanisole and find that both the polarity and hydrogen bonding of the protein scaffold play an important role in tuning the chemoselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T28002
Thioanisole, ReagentPlus®, ≥99%
C7H8S