EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
Bioequivalence study of two oral formulations of cefadroxil in healthy volunteers.
PMID 18368950
Abstract
Two different cefadroxil (CAS 50370-12-2) formulations were evaluated for their relative bioavailability in 24 healthy volunteers who received a single 500 mg oral dose of each preparation. An open, randomized clinical trial designed as a two-period crossover study with a 7-day washout period between doses was employed. Plasma samples for assessments of their cefadroxil concentration by HPLC-UV were obtained over 8 h after administration. Values of 48.94 +/- 10.18 pg x h/ml for test, and 48.51 +/- 9.02 microg x h/ml for the reference preparation AUC(0-t) demonstrate a nearly identical extend of drug absorption. Maximum plasma concentration Cmax of 16.04 +/- 4.94 microg/ml and 16.01 + 4.02 microg/ml achieved for the test and reference preparations did not differ significantly. The parametric 90% confidence intervals (CI) of the mean of the difference (test-reference) between log-transformed values of the two formulations were 96.80% to 104.51% and 92.01% to 107.00% for AUC(0-t) and Cmax, respectively. Since for both AUC(0-t) or Cmax the 90% CI values are within the interval proposed by the Food and Drug Administration, the test product is bioequivalent to the reference product for both the rate and extent of absorption after single dose administration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7020
Cefadroxil, analytical standard
C16H17N3O5S