EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Ground state spin-switching via targeted structural distortion: twisted single-molecule magnets from derivatised salicylaldoximes.
PMID 18369484
Abstract
The use of derivatised salicylaldoximes in Mn chemistry has led to the isolation of a plethora of beautiful new SMMs ranging in nuclearity from three to eight and with spin ground states as large as S = 12-including a Mn6 complex with the largest energy barrier to magnetisation reversal yet reported. The deliberate chemically-induced structural distortion of the [Mn6] molecule allows the isolation of analogous family members displaying remarkably different magnetic properties and this in turn allows for a rare semi-quantitative magneto-structural correlation which enables prediction of the magnetic properties of new family members.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84172
Salicylaldoxime, ≥98.0% (NT)
C7H7NO2