EMAIL THIS PAGE TO A FRIEND
International urology and nephrology
Calciphylaxis: calcific uremic arteriolopathy and the emerging role of sodium thiosulfate.
PMID 18369733
Abstract
Calciphylaxis-calcific uremic arteriolopathy, is a serious disorder of arteriolar calcification of the arteriole media and is associated with endovascular fibrosis and thrombosis in subcutaneous adipose tissue. It frequently results in severe ischemia, intense pain, and tissue necrosis with nonhealing skin ulcerations. It usually occurs in chronic kidney disease and especially in patients requiring renal replacement therapy. It is associated with a very high mortality rate, and the number of reports and reviews seemed to have increased over the past 5 years. Advances in therapy and salvaging patients from this high mortality risk have recently been reported with the use of sodium thiosulfate. The new application for this old drug used to treat cyanide poisoning and recently preventing neurotoxic effects resulting in hearing loss in those patients with head and neck cancer receiving cisplatin and carboplatin therapy are discussed. Recently, multiple case reports have demonstrated that sodium thiosulfate therapy has resulted in rapid pain relief, healing of skin ulcerations, and prevention of high mortality risk. This emerging treatment and its success are relatively unknown to many physicians. The purpose of this report is to share with others the emerging role of sodium thiosulfate and its new application as a treatment option to be used in combination with other treatment modalities for calciphylaxis-calcific uremic arteriolopathy. Indeed, as with any new treatment this emerging therapy should be studied in greater detail, but this old drug seems to have a new life in the hands of treating physicians.
Related Materials