EMAIL THIS PAGE TO A FRIEND
Psychiatria Danubina
Treatment compliance of outpatients with schizophrenia: patient's attitudes, demographic, clinical and therapeutic variables.
PMID 18376330
Abstract
To compare patient's attitudes, demographic, clinical characteristics, psychopathology, insight and type of antipsychotic therapy in compliant and non-compliant outpatients with schizophrenia; to explore correlations between patient's attitudes and related variables. A sample of 44 outpatients of both genders (> 60 years), with a diagnosis of ICD-10 Schizophrenia (F20) was included into the study. All the patients were on maintenance treatment with different classes of antipsychotics (oral, depot or both), for at least 6 months from the latest hospitalisation. The exclusion criteria were determined. The BPRS and the PANSS were used to assess psychopathology and insight (G12 item). The self-report questionnaire MARS was used to assess patient's attitudes. Compliant patients (N=37) showed the following significant differences compared to non-compliant patients (N=7): higher the MARS (p<0.001), lower the PANSS (Positive sub score) (p<0.01) G12 scores (p<0.01) (the Student t test) and percentage of patients with previous non-compliance (p<0.05) (chi2 test). Considerable correlation between the MARS and the BPRS (p<0.001), the PANSS (Positive, General psychopathology) (p<0.001; p<0.01), G12 scores (p<0.05) (negative) and current compliance (p<0.001) was also found (The Spearman's correlation). Our results suggest that special attention should be paid to attitudes, severity of psychopathology, insight and history of non-compliance in compliance evaluation of outpatients with schizophrenia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0260000 Fluphenazine decanoate, European Pharmacopoeia (EP) Reference Standard
C32H44F3N3O2S