EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
The excretion of N1-methyl-2-pyridone-5-carboxylic acid and related compounds in human subjects after oral administration of nicotinic acid, trigonelline and N1-methyl-2-pyridone-5-carboxylic acid.
PMID 1837687
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
741507
1,6-Dihydro-1-methyl-6-oxo-3-pyridinecarboxylic acid, 97%
C7H7NO3