EMAIL THIS PAGE TO A FRIEND
Journal of paediatrics and child health
A favorable response to steroid therapy in a child with drug-associated acute vanishing bile duct syndrome and skin disorder.
PMID 18377375
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7118 Cefdinir, ≥95% (HPLC)
C14H13N5O5S2