EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Solid-phase microextraction method for the determination of volatile compounds associated to oxidation of fish muscle.
PMID 18378245
Abstract
A procedure for the determination of volatile compounds derived from lipid oxidation of fish muscle samples is presented. Analytes are concentrated on a solid-phase microextraction fiber employed in the headspace mode (HS-SPME), and selectively determined using gas chromatography in combination with mass spectrometry (GC-MS). The influence of several parameters on the efficiency of microextraction such as type of fiber, volume of sample, time, temperature, salting-out effect and stirring was systematically investigated. A saline extraction of fish muscle followed by incubation on a Carboxen-polydimethylsiloxane fiber during 30min at 60 degrees C gave the most effective and accurate extraction of the analytes. Quantification of them was performed by MS in the selected ion monitoring mode and by the internal standard method. Satisfactory linearity, repeatability and quantification limits were achieved under these conditions. The method was applied to the determination of the volatile compounds associated to oxidation of Atlantic horse mackerel (Trachurus trachurus) minced muscle and excellent correlations were obtained with chemical indexes for monitoring lipid oxidation as peroxide value and thiobarbituric acid reactive substances. This combined technique is fast, simple, sensitive, inexpensive and useful to monitor target compounds associated to fish rancidity as 1-penten-3-ol, 2,3-pentanedione or 1-octen-3-ol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57318 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, for use with manual holder, needle size 24 ga
57344-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked phase), needle size 23 ga, for use with manual holder
57319 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked), needle size 24 ga, for use with autosampler
57343-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm (partially crosslinked phase), needle size 23 ga, for use with autosampler
57334-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, for use with manual holder, StableFlex fiber
57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, StableFlex, for use with autosampler
57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, StableFlex, for use with autosampler
57906-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm (bonded phase), for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with multi fiber exchanger, needle size 24 ga, StableFlex
504831 SPME Portable Field Sampler, coating Carboxen/Polydimethylsiloxane (CAR/PDMS)