EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Adsorption thermodynamic and kinetic studies of dissolved chromium onto humic acids.
PMID 18378433
Abstract
Adsorption thermodynamic characteristic and kinetics of dissolved chromium from aqueous solutions onto humic acids (HAs) were studied. The results showed that the Freundlich isotherm model described the equilibrium data well. The negative DeltaG degrees confirmed the spontaneity of adsorption process. The positive DeltaH degrees and DeltaS degrees proved the endothermic nature and irreversibility of the sorption. Furthermore, the adsorption process followed the pseudo-second-order equation. With increasing initial concentration of Cr(VI), the adsorption rate constant decreased while the maximum adsorption capacity increased.