EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Simultaneous analysis of 1,2,3,4-tetrahydroisoquinolines by high-performance liquid chromatography using 4-(5,6-dimethoxy-2-phthalimidinyl)-2-methoxyphenylsulfonyl chloride as a fluorescent labeling reagent.
PMID 18378506
Abstract
A highly sensitive high-performance liquid chromatographic method for the simultaneous analysis of 1,2,3,4-tetrahydroisoquinolines (TIQs) in the rat brain was developed. 1,2,3,4-Tetrahydroisoquinoline (TIQ), 1-methyl-1,2,3,4-tetrahydroisoquinoline (1-MeTIQ) and 1-benzyl-1,2,3,4-tetrahydroisoquinoline (1-BeTIQ) were derivatized with 4-(5,6-dimethoxy-2-phthalimidinyl)-2-methoxyphenylsulfonyl chloride to produce fluorescent sulfonamides. The labeling reaction was carried out at 50 degrees C for 15 min at pH 8.5. The fluorescent derivatives were separated on a reversed-phase column by gradient elution using (A) water-(B) acetonitrile/methanol (55:45) at 55 degrees C and detected by fluorescence measurement at 318 nm (excitation) and 398 nm (emission). The detection limits (signal-to-noise ratio=3) were 8-9 fmol per injection. The relative standard deviations (n=6) of TIQs were 2.6-10.5% and the recoveries were 87.6, 101.8 and 75.2%, respectively. The concentrations of TIQ, 1-MeTIQ and 1-BeTIQ in normal rat brains (n=6) were 0.7+/-0.3 (0.10+/-0.04), 3.4+/-1.5 (0.50+/-0.22) and 1.3+/-1.8 pmol/g (0.30+/-0.41 ng/g), respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T13005
1,2,3,4-Tetrahydroisoquinoline, 95%
C9H11N