EMAIL THIS PAGE TO A FRIEND
The European respiratory journal
The biology of oxygen.
PMID 18378783
Abstract
The biology of O(2) is extremely complex and defies a comprehensive yet brief summary that showcases its roles across the entire animal and plant kingdom. This necessarily short introduction to the 2006 Taormina Lung Science Conference (Taormina, Italy) on hypoxia will examine key features of the biology of O(2) only within mammalian cells, even that being a daunting task.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4890 Anti-Human Hemoglobin antibody produced in rabbit, whole antiserum