EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Diels-alder reaction of 2(1H)-pyridones acting as dienes.
PMID 18379094
Abstract
Diels-Alder reactions between N-phenylmaleimide, acting as the dienophile, and 2(1H)-pyridones having a methoxy or a chloro substituent, were carried out, under atmospheric and high pressure conditions, to give the corresponding isoquinuclidine derivatives. Stereoselectivity of the Diels-Alder reactions was studied using molecular orbital calculations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P27100
N-Phenylmaleimide, 97%
C10H7NO2