EMAIL THIS PAGE TO A FRIEND
Ocular immunology and inflammation
Granuloma annulare anterior uveitis.
PMID 18379945
Abstract
To present a case of bilateral granuloma annulare anterior uveitis. Case report. Clinical case presentation and review of literature. A 72-year-old woman with a history of granuloma annulare (GA) presented with acute bilateral granulomatous anterior uveitis. General physical exam revealed hypo- and hyper-pigmented punctate and annular skin lesions on both arms characteristic of GA. Infectious and autoimmune workup revealed a mildly elevated angiotensin converting enzyme (ACE) level. Her uveitis resolved with topical prednisolone and homatropine. Granuloma annulare is a rare cause of anterior uveitis that is diagnosed by characteristic skin findings and may be associated with sarcoidosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0800000
Homatropine hydrobromide, European Pharmacopoeia (EP) Reference Standard
C16H21NO3 · HBr