EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Alkaline Trypan Blue as a stain for superficial fungi.
PMID 18384437
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1144
Chlorazol Black
C34H25N9Na2O7S2