EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Ornicorrugatin, a new siderophore from Pseudomonas fluorescens AF76.
PMID 18386480
Abstract
From a pyoverdin-negative mutant of Pseudomonas fluorescens AF76 a new lipopeptidic siderophore (ornicorrugatin) could be isolated. It is structurally related to the siderophore of Pseudomonas corrugata differing in the replacement of one Dab unit by Orn.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199532
Chromeazurol S, Dye content 50 %
C23H13Cl2Na3O9S