EMAIL THIS PAGE TO A FRIEND
Annales de dermatologie et de venereologie
[Severe toxic dermatitis caused by tenoxicam (Tilcotil). 3 cases].
PMID 1838675
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1762
Isoxicam, analytical standard
C14H13N3O5S