EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact allergy to 1-acetyl-2-phenylhydrazine in a dimethacrylate adhesive.
PMID 1839266
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4626
1-Acetyl-2-phenylhydrazine, ≥98%
C8H10N2O