EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Microwave-assisted efficient copper-promoted N-arylation of amines with arylboronic acids.
PMID 18393467
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517453
Copper(II) acetate, powder, 99.99% trace metals basis
C4H6CuO4
326755
Copper(II) acetate, 98%
C4H6CuO4