EMAIL THIS PAGE TO A FRIEND
Zhonghua yi xue za zhi
[HIV-1 genotypic resistance profiles in children failing highly active antiretroviral therapy].
PMID 18396628
Abstract
To study the genotypic resistance profiles of HIV-1 children failing highly active antiretroviral therapy (HAART) so as to provide helpful information for the treatment regime of Chinese children infected with HIV-1. Peripheral venous blood samples were collected from 20 HIV-1 infected children of Henan province, aged 9 (3 - 14). Nested RT-PCR was used to amplify part of the RT (40 -250 aa) gene. The PCR products of RT gene underwent nucleotide sequencing, the resulting nucleotide sequences were analyzed by the HIVdb data offered by the Stanford University web site to find the drug resistance mutations. (1) Phylogenetic analysis revealed that 20 of the RT sequences were classified as subtype B. (2) According to the genotypic analysis, 20 , 15, and 13 children showed high level resistance to the nevirapine. (NVP), delavirdine (DLV), and efavirenz (EFV) respectively; 7 and 5 children showed high and intermediate level resistance to azidothymidine (AZT) respectively. Five children showed potential low-level and intermediate level resistance to lamivudine (3TC), and 11 showed high level resistance to 3TC; 11 showed intermediate and high level resistance to stavudine (d4T) and didanoside (ddI) respectively; and 19 and 12 children showed resistance to abacavir (ABC) and tenofovir (TDF) which had never been taken by these children. The emergence of HIV resistant strains during antiretroviral therapy is one of the main reasons for treatment failure in HIV-infected children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0180
Delavirdine mesylate, ≥98% (HPLC)
C22H28N6O3S · CH3SO3H