EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of amino-acetonitrile derivatives, a new class of synthetic anthelmintic compounds.
PMID 18400497
Abstract
A new series of amino-acetonitrile derivatives (AAD) have been discovered that exhibit high anthelmintic activity against parasitic nematode species such as Haemonchus contortus and Trichostrongylus colubriformis. Significantly, these compounds also demonstrate activity against nematode strains resistant to the currently available broad-spectrum anthelmintics. The discovery, synthesis, structure-activity relationship and biological results are presented.