EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Development and validation of an LC-MS assay for finasteride and its application to prostate cancer prevention trial sample analysis.
PMID 18402729