EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Time resolved fluorescence spectroscopy of Eu(III) complexation with alpha-hydroxy isobutyric acid.
PMID 18406663
Abstract
Complexation of Eu(III) with alpha-hydroxy isobutyric acid (HIBA), a model compound of humic acid, has been studied by time resolved fluorescence spectroscopy. The ratio of fluorescence intensity of the two peaks at 616 and 592 nm (I(616/592)) was found to increase with increasing ligand to metal ratio. The I(616/592) data was used to deduce the stability constant of Eu-HIBA complexes of the type ML(i) (i=1-3). The formation of multiple ligand complexes was also corroborated by lifetime data which was found to increase with increasing [HIBA]/[Eu] ratio thus indicating replacement of coordinated water molecules by HIBA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323594
α-Hydroxyisobutyric acid, 99%
C4H8O3
164976
α-Hydroxyisobutyric acid, 98%
C4H8O3