EMAIL THIS PAGE TO A FRIEND
Natural product research
Biotransformation of tetrahydro-alpha-santonins by Absidia coerulea.
PMID 18415857
Abstract
The fungus, Absidia coerulea was employed to bioconvert tetrahydro-alpha-santonins, 1,2,4alpha,5alpha-tetrahydro-alpha-santonin (1), and its 4-epimer (2), from which 10 products (3-12) were obtained. Furthermore, their structures were determined, based on their chemical and spectroscopic data analyses. Among them, 3-5, 7, 9, 11 and 12 were observed to be seven new compounds. The reactions mainly involved in these bio-process included hydroxylation(s) (C-4, C-11, and C-1), reduction (C-3 ketone to alcohol).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1250
(−)-α-Santonin, ≥99%
C15H18O3
53653
(−)-α-Santonin, analytical standard
C15H18O3