EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Proliferation of L02 human hepatocytes in tolerized genetically immunocompetent rats.
PMID 18416458
Abstract
To investigate whether human hepatocytes could proliferate after transplantation to normal immunocompetent rats treated with 2-acetaminofluorene or retrorsine and partial hepatectomy. L02 hepatocyte-tolerant Sprague-Dawley rats were injected with retrorsine, 2-acetaminofluorene or normal saline. L02 hepatocytes were then transplanted via the spleen. Human albumin and its mRNA, specific proliferating cell nuclear antigen (PCNA), L02 hepatocyte dynamic distribution, number density and area density of PCNA-positive cells in the liver were determined. All the examined indicators were not significantly different between the rats treated with 2-acetaminofluorene and normal saline, which was not the case with rats treated with retrorsine. A dynamic distribution of L02 hepatocytes in the rat liver was detected from wk 1 to mo 6 after transplantation in the retrorsine group and from wk 1 to 10 in the 2-acetaminofluorene group. Human albumin and its mRNA were detected from wk 2 to mo 6 in the retrorsine group and from wk 1 to 8 in the 2-acetaminofluorene group. Specific human PCNA was detected in the rat liver from wk 2 to mo 6 in the retrorsine group and from wk 2 to 6 in the 2-acetaminofluorene group. Human albumin and its mRNA contents as well as the number of PCNA positive cells reached a peak at wk 4. L02 human hepatocytes could not proliferate significiantly after transplantation to the normal, immunocompetent rats treated with 2-acetaminofluorene. L02 human hepatocytes can survive for 10 wk after transplantation and express human albumin for 8 wk. L02 human hepatocytes can proliferate and express human albumin for 6 mo after transplantation to the rats treated with retrorsine. The chimeric L02 human hepatocytes, which then underwent transplantation into tolerant rats, were normal in morphogenesis, biochemistry and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0382
Retrorsine, ≥90% (HPLC)
C18H25NO6
37025
Retrorsine, analytical standard
C18H25NO6