EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Determination of fluoxetine in plasma by gas chromatography-mass spectrometry using stir bar sorptive extraction.
PMID 18420052
Abstract
This article presents a method employing stir bar sorptive extraction (SBSE) with in situ derivatization, in combination with either thermal or liquid desorption on-line coupled to gas chromatography-mass spectrometry for the analysis of fluoxetine in plasma samples. Ethyl chloroformate was employed as derivatizing agent producing symmetrical peaks. Parameters such as solvent polarity, time for analyte desorption, and extraction time, were evaluated. During the validation process, the developed method presented specificity, linearity (R(2)>0.99), precision (R.S.D.<15%), and limits of quantification (LOQ) of 30 and 1.37 pg mL(-1), when liquid and thermal desorption were employed, respectively. This simple and highly sensitive method showed to be adequate for the measurement of fluoxetine in typical and trace concentration levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185892
Ethyl chloroformate, 97%
C3H5ClO2