EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Clinical practice. Irritable bowel syndrome.
PMID 18420501
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0346
Alosetron hydrochloride, ≥98% (HPLC)
C17H18N4O·HCl