EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A modular flow reactor for performing Curtius rearrangements as a continuous flow process.
PMID 18421389
Abstract
The use of a mesofluidic flow reactor is described for performing Curtius rearrangement reactions of carboxylic acids in the presence of diphenylphosphoryl azide and trapping of the intermediate isocyanates with various nucleophiles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178756
Diphenyl phosphoryl azide, 97%
C12H10N3O3P