EMAIL THIS PAGE TO A FRIEND
Experimental neurology
A dopamine receptor antagonist L-745,870 suppresses microglia activation in spinal cord and mitigates the progression in ALS model mice.
PMID 18423451