EMAIL THIS PAGE TO A FRIEND
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP)
The ratio of lithocholic to deoxycholic acid in faeces: a risk factor in colorectal carcinogenesis.
PMID 1842737