EMAIL THIS PAGE TO A FRIEND
Archiwum medycyny sadowej i kryminologii
[Poisonings with anticoagulant rodenticides].
PMID 18432144
Abstract
Anticoagulant rodenticides are commonly used in extermination of rodents. Failure to adhere to safety principles and sometimes a deliberate use of these compounds may be a reason of severe poisonings in human. Diagnosis is based on a characteristic clinical course of the disease and postmortem examinations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32335
Bromethalin, PESTANAL®, analytical standard
C14H7Br3F3N3O4