EMAIL THIS PAGE TO A FRIEND
Archiwum medycyny sadowej i kryminologii
[Suspected azodicarbonamide poisoning in a patient with acute hemorrhaging pancreatitis].
PMID 18432145
Abstract
The report describes the case of death of a 22-year old man due to acute hemorrhaging pancreatitis. Azodicarbonamide poisoning was suspected in this patient. Autopsy results, data from the literature and the knowledge about mechanisms of azodicarbonamide toxicity allowed for excluding poisoning with this substance as the cause of death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A96606
Azodicarboxamide, 97%
C2H4N4O2