EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric total synthesis of antiochic acid.
PMID 18439020
Abstract
The first asymmetric total synthesis of antiochic acid using bioinspired polyene cyclization strategy is described. Both good yield and good asymmetric induction were obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
663212
Isopropenylboronic acid pinacol ester, contains phenothiazine as stabilizer, 95%
C9H17BO2