EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
VIP and VIP gene silencing modulation of differentiation marker N-cadherin and cell shape of corneal endothelium in human corneas ex vivo.
PMID 18441300
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-729
Anti-Bcl2 Antibody, clone 100, clone 100, Upstate®, from mouse