EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Catalytic asymmetric Michael reactions of acetaldehyde.
PMID 18442029
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N26806
trans-β-Nitrostyrene, 99%
C8H7NO2