EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Tuning of the H-transfer coordinate in primitive versus well-evolved enzymes.
PMID 18444258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7006
Dihydrofolic acid, ≥90%
C19H21N7O6