EMAIL THIS PAGE TO A FRIEND
Journal of microscopy
Microcarriers for high-pressure freezing and cryosectioning of adherent cells.
PMID 18445159
Abstract
A method is described employing microcarrier spheres of cross-linked dextran for obtaining ultra- and semithin vitreous sections from high-pressure frozen anchorage-dependent (mammalian) cells. Avoiding trypsination or scraping cells off from the culture surface, the presented approach allows for cryoimmobilization, cryosectioning and cryoelectron microscopy/tomography of frozen-hydrated cells in an unperturbed manner which is important to preserve the native state of, for instance, the cytoskeleton. Furthermore, our studies on the 'life cycle' of Herpes simplex virus in Vero cells demonstrate that cell monolayers on microcarrier beads are well suited for fluorescence microscopic characterization of the sample prior to high-pressure freezing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0646 Cytodex® 1 microcarrier beads, Cytodex 1, 60-87 μm